Merck
  • Intracellular localization and partial amino acid sequence of a stress-inducible 40-kDa protein in HeLa cells.
Intracellular localization and partial amino acid sequence of a stress-inducible 40-kDa protein in HeLa cells.
Cell structure and function (1992-02-01)
H Hattori, Y C Liu, I Tohnai, M Ueda, T Kaneda, T Kobayashi, K Tanabe, K Ohtsuka
ABSTRACT
We earlier discovered a novel 40-kDa protein (hsp40) induced by heat shock and other stresses in mammalian and avian cells. In this report, we purified the hsp40 in HeLa cells, using modified two-dimensional gel electrophoresis, and determined the amino terminal amino acid sequence of this protein. The hsp40 is homologous to DnaJ, an Escherichia coli heat-shock protein, as well as to DnaJ-homologous proteins in yeast such as SCJ1, Sec63/Np11, YDJ1 and SIS1. Indirect immunofluorescence staining using an anti-hsp40 polyclonal antibody demonstrated that hsp40 was localized faintly throughout the cell in non-heat-shocked cells and was accumulated in nuclei and nucleoli in heat-shocked cells. The intracellular localization of hsp40 was very similar to that of hsp70, suggesting that these two hsps colocalize in heat-shocked HeLa cells.
Sigma-Aldrich
HSP40, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution