Merck
  • Differential dopaminergic regulation of proenkephalin and prodynorphin mRNAs in the basal ganglia of rats.
Differential dopaminergic regulation of proenkephalin and prodynorphin mRNAs in the basal ganglia of rats.
Brain research. Molecular brain research (1992-01-01)
S Reimer, D J Sirinathsinghji, K E Nikolorakis, V Höllt
ABSTRACT
Proenkephalin and prodynorphin mRNA levels in the caudate-putamen and in the nucleus accumbens of rats were measured by in situ hybridization 2, 4 and 8 weeks following unilateral lesion of the medial forebrain bundle by 6-hydroxydopamine (6-OHDA). After 2 weeks a 60% increase of the levels of proenkephalin mRNA in the ipsilateral caudate-putamen was observed which declined to 20% above control after 8 weeks. A smaller increase in the levels of proenkephalin mRNA of about 20% was observed in the nucleus accumbens after 2 weeks and no significant alteration could be observed 4 and 8 weeks after lesioning. The levels of prodynorphin mRNA in the ipsilateral caudate-putamen decreased 20% below control and returned to control levels 4 and 8 weeks post-lesion. In contrast, in the nucleus accumbens a persistent ipsilateral decrease of prodynorphin (20-30%) was found 2, 4 and 8 weeks post-lesion. These findings indicate, that lesions of the mesostriatal dopamine (DA) system differentially influences opioidergic gene expression in distinct areas of the caudate-putamen. The lesions cause an increase in proenkephalin mRNA levels which was higher in the caudate-putamen than in the nucleus accumbens and tend to be reversible. Conversely, the lesion caused a persistent decrease in the levels of prodynorphin mRNA in the nucleus accumbens and a small and transient decrease in the caudate-putamen.
Sigma-Aldrich
5-Hydroxydopamine hydrochloride, 98%