Merck
  • Environmental effects and desorption characteristics on heavy metal removal using carboxylated alginic acid.
Environmental effects and desorption characteristics on heavy metal removal using carboxylated alginic acid.
Bioresource technology (2004-09-15)
Choong Jeon, Young Je Yoo, Wolfgang H Hoell
ABSTRACT
Effects of ionic strength and organic materials on copper ion uptake capacity using carboxylated alginic acid which showed very high metal ion uptake capacity were investigated. The ionic strength only had a slight effect on the decrease of copper ion uptake capacity regardless of NaCl concentration. And, the effect of organic materials such as NTA (nitrilotriaceticacid) and sodium hypophosphite on the copper ion uptake capacity was negligible. When the lead ion adsorbed on carboxylated alginic acid was desorbed by NTA, which showed high desorption efficiency, the best optimum concentration of NTA was about 0.01 M. Also desorption efficiency decreased, however, concentration factor increased as S/L ratio which is defined as the ratio of adding amount of adsorbed and volume of desorbing agent increased. In sequential adsorption and desorption cycles, the lead uptake capacity on carboxylated alginic acid was relatively maintained through cycles 1-5.
Sigma-Aldrich
Sodium hypophosphite monohydrate, ≥99%
Sigma-Aldrich
Sodium hypophosphite monohydrate, puriss. p.a., reag. Ph. Eur., ≥99%
Sigma-Aldrich
Sodium hypophosphite, puriss., meets analytical specification of BPC63, anhydrous, 98-101%