Merck
  • Monodentate phosphines provide highly active catalysts for Pd-catalyzed C-N bond-forming reactions of heteroaromatic halides/amines and (H)N-heterocycles.
Monodentate phosphines provide highly active catalysts for Pd-catalyzed C-N bond-forming reactions of heteroaromatic halides/amines and (H)N-heterocycles.
Angewandte Chemie (International ed. in English) (2006-09-07)
Kevin W Anderson, Rachel E Tundel, Takashi Ikawa, Ryan A Altman, Stephen L Buchwald
Sigma-Aldrich
Hexylamine, 99%