Merck
  • Pyrones to pyrans: enantioselective radical additions to acyloxy pyrones.
Pyrones to pyrans: enantioselective radical additions to acyloxy pyrones.
Journal of the American Chemical Society (2006-10-13)
Mukund P Sibi, Jake Zimmerman
ABSTRACT
This paper describes a highly site-, diastereo-, and enantioselective intermolecular radical addition/hydrogen atom transfer to hydroxypyrone pyromeconic and kojic acids. The methodology can be extended to the formation of chiral quaternary centers. The products obtained are densely functionalized pyran moieties. The products contain structural features amenable for the introduction of additional substituents.
Sigma-Aldrich
1,2,4,5-Benzenetetracarboxylic acid, 96%
Sigma-Aldrich
Pivalic acid, 99%