Merck
  • Extracellular terbium and divalent cation effects on the red blood cell Na pump and chrysoidine effects on the renal Na pump.
Extracellular terbium and divalent cation effects on the red blood cell Na pump and chrysoidine effects on the renal Na pump.
Blood cells, molecules & diseases (2007-04-27)
Matthew S Reifenberger, Krista L Arnett, Craig Gatto, Mark A Milanick
ABSTRACT
We examined the effect of extracellular terbium (Tb(3+)) and divalent metal cations (Ca(2+), Sr(2+), and Ba(2+)) on (86)Rb(+) influx into rabbit and human red blood cells. We found that Tb(3+) at 15 and 25 microM was a non-competitive inhibitor of (86)Rb(+) influx suggesting that Tb(3+) is not binding to the transport site. This result reduces the usefulness of Tb(3+) as a potential probe for the E(out) conformation (the conformation with the transport site facing extracellularly). Ba(2+), Sr(2+) and Ca(2+), at concentrations >50 mM, had minimal effects on Rb(+) influx into red blood cells (1 mM Rb-out). This suggests that the outside transport site is very specific for monovalent cations over divalent cations, in contrast to the inside transport site. We also found that chrysoidine (4-phenylazo-m-phenylenediamine) competes with Na(+) for ATPase activity and K(+) for pNPPase activity suggesting it is binding to the E(in) conformation. Chrysoidine and similar compounds may be useful as optical probes of the E(in) conformation.
Sigma-Aldrich
Chrysoidine G, suitable for microscopy
Supelco
Chrysoidine G, analytical standard