Merck
  • Formation of hydrophobic bridges between multicompartment micelles of miktoarm star terpolymers in water.
Formation of hydrophobic bridges between multicompartment micelles of miktoarm star terpolymers in water.
Chemical communications (Cambridge, England) (2009-02-20)
Andreas Walther, Axel H E Müller
ABSTRACT
We report on the unexpected finding of the formation of hydrophobic bridges between polybutadiene patches of multicompartment micelles based on amphiphilic miktoarm star terpolymers in water.
Product Description
Supelco
Poly(2-vinylpyridine), analytical standard, average Mw 5,000 (Typical), average Mn 4,800 (Typical)
Supelco
Poly(2-vinylpyridine), analytical standard, average Mw 37,500 (Typical), average Mn 35,000 (Typical)
Supelco
Poly(2-vinylpyridine), analytical standard, average Mw 159,000 (Typical), average Mn 152,000 (Typical)