Merck
Photolytic removal and mineralisation of 2-halogenated pyridines.
Water research (2009-07-14)
David R Stapleton, Ioannis K Konstantinou, Dimitra G Hela, Maria Papadaki
ABSTRACT
The photolytic destruction and mineralisation of 2-halogenated pyridines (2-HalPYs) was studied in a range of conditions by means of ultraviolet irradiation at 254 nm. In all cases pseudo-first order kinetics satisfactorily describe the degradation process. Key decomposition products were identified and primary reaction pathways are proposed. 2-HalPYs were rapidly dehalogenated with a subsequent sharp pH drop. Complete Total Organic Carbon (TOC) removal was achieved. Their photolytic removal rate is not affected by pH or aeration. The fastest TOC removal was observed in 2-fluoropyridine (2-FPY) photolysis. 2-Hydroxypyridine (2-HPY) was identified as the primary intermediate formed by destruction of 2-chloropyridine (2-CPY), 2-bromopyridine (2-BPY) and 2-iodopyridine (2-IPY). 2-HPY is further destructed to form Dewar pyridinone. 2-FPY only marginally reacted to form 2-HPY. Instead, its major intermediate product is Dewar pyridinone. 2-IPY results in a, most likely aliphatic, photostable product, the production of which appears to increase at low pH and high 2-HPY concentration.
Sigma-Aldrich
2-Chloropyridine, 99%
Pricing and availability is not currently available.