Merck
  • [Development of anti-trypanosome drugs targeting nucleotides biosynthesis and red-ox regulatory pathway].
[Development of anti-trypanosome drugs targeting nucleotides biosynthesis and red-ox regulatory pathway].
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme (2009-09-01)
Kiyoshi Kita, Daniel Ken Inaoka, Shigeharu Harada
Sigma-Aldrich
L-Dihydroorotic acid, ≥99%
Sigma-Aldrich
DL-Dihydroorotic acid
Sigma-Aldrich
D-Hydroorotic acid, 98%