Merck
  • The spin trapping of superoxide with M4PO (3,3,5,5-tetramethylpyrroline-N-oxide).
The spin trapping of superoxide with M4PO (3,3,5,5-tetramethylpyrroline-N-oxide).
Free radical biology & medicine (1990-01-01)
G R Buettner, B E Britigan
ABSTRACT
The spin trap 3,3,5,5-tetramethylpyrroline-N-oxide (M4PO) reacts with superoxide to produce a short-lived spin adduct (M4PO/.OOH, AN = 14.0 G, AH = 6.5 G) that decays by a first-order process (t1/2 approximately 35 s at pH 7.4). This short lifetime may limit its usefulness in studies of superoxide.
Sigma-Aldrich
3,3,5,5-Tetramethyl-1-pyrroline N-oxide, 95%