Merck
  • Solanum nigrum L. weed plants as a remediation tool for metalaxyl-polluted effluents and soils.
Solanum nigrum L. weed plants as a remediation tool for metalaxyl-polluted effluents and soils.
Chemosphere (2011-07-12)
Jorge Teixeira, Alexandra de Sousa, Manuel Azenha, José Tiago Moreira, Fernanda Fidalgo, A Fernando Silva, Joaquim L Faria, Adrián M T Silva
ABSTRACT
In this work, the phytoremediation potential of metalaxyl, a commonly used persistent, mobile and leachy fungicide, by Solanum nigrum L. plants was studied. The study revealed that this plant species can be used as an excellent metalaxyl phytoremediation tool, thus providing a cost effective and environmentally friendly clean technology for the decontamination of sites and effluents. As it can be sowed directly in the remediation site, is able to complete its life cycle without suffering major stress. Because it accumulates high amounts of the fungicide in the aboveground tissues, enables its concentration and proper disposal by cutting off the corresponding plant part. The study also suggests that the tolerance to metalaxyl is due to a suitable antioxidant response comprising proline accumulation and guaiacol peroxidase and glutathione-S-transferase enhanced activities, that reduce oxidative damage to the plant organs.
Product Description
Metalaxyl, analytical standard
Supelco
Metalaxyl, PESTANAL®, analytical standard