Merck
  • Homogeneous vs heterogeneous polymerization catalysis revealed by single-particle fluorescence microscopy.
Homogeneous vs heterogeneous polymerization catalysis revealed by single-particle fluorescence microscopy.
Journal of the American Chemical Society (2011-10-20)
N Melody Esfandiari, Suzanne A Blum
ABSTRACT
A high-sensitivity and high-resolution single-particle fluorescence microscopy technique differentiated between homogeneous and heterogeneous metathesis polymerization catalysis by imaging the location of the early stages of polymerization. By imaging single polymers and single crystals of Grubbs II, polymerization catalysis was revealed to be solely homogeneous rather than heterogeneous or both.
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer