Merck
  • Chemical constituents comparison between Rhizoma Smilacis Glabrae and Rhizoma Smilacis Chinae by HPLC-DAD-MS/MS.
Chemical constituents comparison between Rhizoma Smilacis Glabrae and Rhizoma Smilacis Chinae by HPLC-DAD-MS/MS.
Natural product research (2012-03-10)
Qing-Feng Zhang, Yu-Xian Guo, Guodong Zheng, Wen-Jun Wang
ABSTRACT
Rhizoma Smilacis Glabrae (RSG) and Rhizoma Smilacis Chinae (RSC) are two herbal materials that belong to the same genera and are both listed in the Chinese Pharmacopoeia. Chemical constituents in the two species were compared by HPLC-DAD-MS/MS. Many common constituents were found in both species, including shikimic acid, 5-O-caffeoylshikimic acid, trans-resveratrol, taxifolin, astilbin and its three stereoisomers, engeletin and isoengeletin. However, syringic acid was found only in RSG, while chlorogenic acid was found only in RSC.
Product Description
Supelco
Taxifolin, analytical standard
Sigma-Aldrich
Astilbin from Engelhardtia roxburghiana, ≥98%