Merck
  • Abietane diterpenes induce cytotoxic effects in human pancreatic cancer cell line MIA PaCa-2 through different modes of action.
Abietane diterpenes induce cytotoxic effects in human pancreatic cancer cell line MIA PaCa-2 through different modes of action.
Phytochemistry (2012-03-23)
Marcio Fronza, Evelyn Lamy, Stefan Günther, Berta Heinzmann, Stefan Laufer, Irmgard Merfort
ABSTRACT
Abietane diterpenes, especially those containing quinone moieties, are often reported to have cytotoxic effects on cancer cell lines. They deserve greater attention because several cancer chemotherapeutic agents also possess the quinone structural feature. To date, very little is known about their cytotoxic molecular modes of action. In the present study, five diterpenes, 7 alpha-acetoxyroyleanone, horminone, royleanone, 7-ketoroyleanone and sugiol which have been previously isolated from the medicinal plant Peltodon longipes were shown to possess cytotoxic activity against the human pancreatic cancer cell line MIA PaCa-2. 7 alpha-Acetoxyroyleanone, horminone and royleanone were demonstrated to possess alkylating properties using the nucleophile 4-(4-nitrobenzyl)pyridine. However, no clear correlation between the alkylating properties and cytotoxicity of these diterpenes was observed. Furthermore, the relaxation activity of human DNA topoisomerases I and II was found to be influenced by these compounds, with 7-ketoroyleanone and sugiol being the most active. These two diterpenes preferentially inhibited topoisomerase I and exhibited lower IC(50) values than the classical topoisomerase I inhibitor camptothecin. Molecular docking studies revealed possible interactions of diterpenes with topoisomerase I, indicating that these compounds do not form the drug-enzyme-DNA covalent ternary complex as observed with camptothecin. A binding pocket located at the surface of the DNA-interaction site was proposed. Moreover, the ability of the five diterpenes to generate DNA-strand breaks in single cells was confirmed using the alkaline comet assay. As expected, these diterpenes also influenced cell cycle progression and arrested cells in different phases of the cell cycle, primarily the G1/G0 and S-phases. Interestingly, the diterpenes only exhibited a slight ability to induce apoptotic cell death and failed to generate intracellular reactive oxygen species. These results provide additional understanding of the cytotoxic effects of abietane diterpenes. Depending on their functional groups, we propose that abietane diterpenes utilise different mechanisms to induce cell death.
Sigma-Aldrich
4-(4-Nitrobenzyl)pyridine, 98%