Merck
Stability of aflatoxins in solution.
Journal of AOAC International (2012-09-14)
Gonzalo J Diaz, Sandra M Cepeda, Perry A Martos
ABSTRACT
The stability of aflatoxins B1, B2, G1, and G2 was studied in solutions containing different concentrations of water, acetonitrile, and/or methanol, and in autosampler vials treated with nitric acid or silanized. When stored at room temperature (20 degrees C) for 24 h, aflatoxins G1 and G2 were stable only in solutions containing 100% organic solvent, whereas aflatoxins B1 and B2 were stable in solutions of methanol-water and acetonitrile-water at greater than 60 and 40% organic content, respectively. At 5 degrees C, aflatoxins G1 and G2 showed a significant decrease in concentration only when kept in less than 20% aqueous organic solvent. Significant loss of aflatoxins was realized in standard, commercially available amber type I borosilicate autosampler vials, but chemical etching of the vials with nitric acid or with silanization prevented aflatoxin degradation. These results indicate that aflatoxins are unstable in aqueous solutions and that this instability can be counteracted by the presence of at least 20% organic solvent and keeping the solutions at 5 degrees C or by the use of treated vials.
Sigma-Aldrich
Aflatoxin B2
Supelco
Aflatoxin G2 solution, 0.5 μg/mL in acetonitrile, analytical standard
Aflatoxin G2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B2 solution, 0.5 μg/mL in acetonitrile, analytical standard
Aflatoxin G1 solution, 3.78 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin G1 solution, 2 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin B2
Aflatoxin B2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Aflatoxin G2
Sigma-Aldrich
Nitric acid, reagent grade, fuming, >90%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Supelco
Aflatoxin G2
Supelco
Aflatoxin G1
Sigma-Aldrich
Aflatoxin G1, from Aspergillus flavus
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, purum p.a., fuming, packed in coated, shock- and leak-protected glass bottle, ≥99% (T)
Sigma-Aldrich
Nitric acid, ACS reagent, 70%