Merck
  • Nano-positioning system for structural analysis of functional homomeric proteins in multiple conformations.
Nano-positioning system for structural analysis of functional homomeric proteins in multiple conformations.
Structure (London, England : 1993) (2012-10-16)
H Clark Hyde, Walter Sandtner, Ernesto Vargas, Alper T Dagcan, Janice L Robertson, Benoit Roux, Ana M Correa, Francisco Bezanilla
ABSTRACT
Proteins may undergo multiple conformational changes required for their function. One strategy used to estimate target-site positions in unknown structural conformations involves single-pair resonance energy transfer (RET) distance measurements. However, interpretation of inter-residue distances is difficult when applied to three-dimensional structural rearrangements, especially in homomeric systems. We developed a positioning method using inverse trilateration/triangulation to map target sites within a homomeric protein in all defined states, with simultaneous functional recordings. The procedure accounts for probe diffusion to accurately determine the three-dimensional position and confidence region of lanthanide LRET donors attached to a target site (one per subunit), relative to a single fluorescent acceptor placed in a static site. As first application, the method is used to determine the position of a functional voltage-gated potassium channel's voltage sensor. Our results verify the crystal structure relaxed conformation and report on the resting and active conformations for which crystal structures are not available.
Sigma-Aldrich
Terbium, foil, size 25 mm × 25 mm × 1 mm, 99.9% trace rare earth metals basis