Merck
  • A facile, relative green, and inexpensive synthetic approach toward large-scale production of SnS₂ nanoplates for high-performance lithium-ion batteries.
A facile, relative green, and inexpensive synthetic approach toward large-scale production of SnS₂ nanoplates for high-performance lithium-ion batteries.
Nanoscale (2013-01-12)
Yaping Du, Zongyou Yin, Xianhong Rui, Zhiyuan Zeng, Xue-Jun Wu, Juqing Liu, Yuanyuan Zhu, Jixin Zhu, Xiao Huang, Qingyu Yan, Hua Zhang
ABSTRACT
A facile, environmentally friendly, and economical synthetic route for production of large-amounts (gram scale) of two-dimensional (2D) layered SnS(2) nanoplates is presented. The electrode fabricated from the SnS(2) nanoplate exhibits excellent lithium-ion battery performance with highly reversible capacity, good cycling stability and excellent capacity retention after 30 cycles.
Sigma-Aldrich
Tin(II) sulfide, 96%
Sigma-Aldrich
Tin(II) sulfide, granular, ≥99.99% trace metals basis