- Evaluation of the anti-inflammatory effect of chalcone and chalcone analogues in a zebrafish model.
Evaluation of the anti-inflammatory effect of chalcone and chalcone analogues in a zebrafish model.
Molecules (Basel, Switzerland) (2013-02-07)
Yau-Hung Chen, Wei-Hua Wang, Yun-Hsin Wang, Zi-Yu Lin, Chi-Chung Wen, Ching-Yuh Chern
PMID23385341
ABSTRACT
The aim of this study was to investigate novel chalcones with potent anti-inflammatory activities in vivo. Chalcone and two chalcone analogues (compound 5 and 9) were evaluated using a caudal fin-wounded transgenic zebrafish line "Tg(mpx:gfp)" to visualize the effect of neutrophil recruitment dynamically. Results showed that treatment with compound 9 not only affected wound-induced neutrophil recruitment, but also affected Mpx enzymatic activity. Moreover, protein expression levels of pro-inflammatory factors (Mpx, NFκB, and TNFα) were also regulated by compound 9. Taken together, our results provide in vivo evidence of the anti-inflammatory effects of synthesized chalcone analogues on wound-induced inflammation.
MATERIALS