Merck
  • N-benzyladriamycin-14-valerate versus progressively doxorubicin-resistant murine tumours: cellular pharmacology and characterisation of cross-resistance in vitro and in vivo.
N-benzyladriamycin-14-valerate versus progressively doxorubicin-resistant murine tumours: cellular pharmacology and characterisation of cross-resistance in vitro and in vivo.
British journal of cancer (1989-12-01)
R Ganapathi, D Grabowski, T W Sweatman, R Seshadri, M Israel
ABSTRACT
N-Benzyladriamycin-14-valerate (AD198) is a novel lipophilic anthracycline with greater in vivo antitumour activity than doxorubicin (DOX) in experimental model systems. Using sensitive and progressively DOX-resistant L1210 mouse leukaemia and B16-BL6 mouse melanoma lines, we have determined the cellular pharmacokinetics and cytotoxic response in vitro and in vivo of AD198. In the L1210 leukaemia model following 3 h drug exposure in vitro, the IC50 for AD198 was approximately 0.35 microgram ml-1 for the sensitive and 10-fold DOX resistant cells and 1.0 microgram ml-1 for the 40-fold DOX resistant cells. A similar pattern of cross-resistance to AD198 was also observed with the B16-BL6 melanoma, with and IC50 for AD198 with the sensitive and 10-fold DOX-resistant cells being similar, and about 2-fold higher with the 40-fold resistant cells. In the L1210 leukaemia model, cellular pharmacokinetics of AD198 revealed the following: (a) accumulation of AD198 was concentration but not time dependent, and cellular drug levels in the sensitive and resistant sublines were similar when treated with equimolar concentrations; (b) retention of AD 198 was 60% of the initial drug uptake and, in cells treated with the IC50 of AD198, cellular levels in the 40-fold DOX-resistant line were, as expected, 2-fold higher than in sensitive or 10-fold DOX-resistant cells; (c) in vitro biotransformation of AD 198 in the sensitive and resistant sublines was comparable. Studies in vivo with i.p. L1210 leukaemia (disseminating) and B16-BL6 melanoma (non-disseminating) tumour models evaluating therapeutic efficacy of DOX vs AD 198 in mice implanted with tumour i.p. on day 0 and treated i.p. on days 1-4 indicated: (a) DOX at 3 mg kg-1 administered once daily on days 1-4 resulted in a 55% ILS and 104% ILS with parent-sensitive B16-BL6 melanoma and L1210 leukaemia models respectively; however, similar doses of DOX in the resistant sublines were ineffective, with survival similar to the untreated control; (b) AD198 at 10-12.5 mg kg-1 day-1 for 4 days was extremely effective in the sensitive L1210 (189% ILS), and similar to DOX (61% ILS) in the sensitive B16-BL6; (c) AD198 (10-12.5 mg kg-1) was ineffective (survival similar to untreated control) in the 10-and 40-fold DOX-resistant L1210 leukaemia and 40-fold DOX resistant B16-BL6 melanoma, but produced a 76% ILS in the 10-fold DOX resistant B16-BL6 melanoma.(ABSTRACT TRUNCATED AT 400 WORDS)