Merck
  • Direct electron spin resonance detection of free radical intermediates during the peroxidase catalyzed oxidation of phenacetin metabolites.
Direct electron spin resonance detection of free radical intermediates during the peroxidase catalyzed oxidation of phenacetin metabolites.
Chemico-biological interactions (1986-11-01)
V Fischer, L S Harman, P R West, R P Mason
ABSTRACT
The oxidation of the phenacetin metabolites p-phenetidine and acetaminophen by peroxidases was investigated. Free radical intermediates from both metabolites were detected using fast-flow ESR spectroscopy. Oxidation of acetaminophen with either lactoperoxidase and hydrogen peroxide or horseradish peroxidase and hydrogen peroxide resulted in the formation of the N-acetyl-4-aminophenoxyl free radical. Totally resolved spectra were obtained and completely analyzed. The radical concentration was dependent on the square root of the enzyme concentration, indicating second-order decay of the radical, as is consistent with its dimerization or disproportionation. The horseradish peroxidase/hydrogen peroxide-catalyzed oxidation of p-phenetidine (4-ethoxyaniline) at pH 7.5-8.5 resulted in the one-electron oxidation products, the 4-ethoxyaniline cation free radical. The ESR spectra were well resolved and could be unambiguously assigned. Again, the enzyme dependence of the radical concentration indicated a second-order decay. The ESR spectrum of the conjugate base of the 4-ethoxyaniline cation radical, the neutral 4-ethoxyphenazyl free radical, was obtained at pH 11-12 by the oxidation of p-phenetidine with potassium permanganate.
Sigma-Aldrich
4-Ethoxyaniline, 98%
Pricing and availability is not currently available.