Merck
  • Production of S-(+)-2-phenylpropionic acid from (R,S)-2-phenylpropionitrile by the combination of nitrile hydratase and stereoselective amidase in Rhodococcus equi TG328.
Production of S-(+)-2-phenylpropionic acid from (R,S)-2-phenylpropionitrile by the combination of nitrile hydratase and stereoselective amidase in Rhodococcus equi TG328.
Applied microbiology and biotechnology (1993-08-01)
T Gilligan, H Yamada, T Nagasawa
ABSTRACT
A new soil isolate, tentatively identified as Rhodococcus equi TG328, was found to be effective in the production of S-(+)-2-phenylpropionic acid from (R,S)-2-phenylpropionitrile. The conversion is catalysed by two enzymes. First, a nitrile hydratase converts the (R,S)-nitrile to (R,S)-2-phenylpropionamide. Second, a stereoselective amidase converts the S-(+)-amide to S-(+)-2-phenylpropionic acid. Conditions for optimal enzyme production and accumulation of S-(+)-2-phenylpropionic acid by resting cells were studied. The reaction of resting cells for 30 h at 10 degrees C with (R,S)-2-phenylpropionitrile resulted in the production of 100 g of S-(+)-2-phenylpropionic acid per litre of reaction mixture. The enantiometric excess of the purified S-(+)-2-phenylpropionic acid was 99.4%. The amount of S-(+)-2-phenylpropionic acid accumulated was enhanced by lower reaction temperatures. In addition, unreacted R-(-)-2-phenylpropionamide with 99.0% enantiometric excess was isolated.
Sigma-Aldrich
α-Methylbenzyl cyanide, 96%
Sigma-Aldrich
(R)-(−)-2-Phenylpropionic acid, 97%
Sigma-Aldrich
(S)-(+)-2-Phenylpropionic acid, 97%
Sigma-Aldrich
2-Phenylpropionic acid, 97%