Merck
  • Assignment of the 750 MHz 1H NMR resonances from a mixture of transacylated ester glucuronic acid conjugates with the aid of oversampling and digital filtering during acquisition.
Assignment of the 750 MHz 1H NMR resonances from a mixture of transacylated ester glucuronic acid conjugates with the aid of oversampling and digital filtering during acquisition.
Journal of pharmaceutical and biomedical analysis (1995-07-01)
R D Farrant, M Spraul, I D Wilson, A W Nicholls, J K Nicholson, J C Lindon
ABSTRACT
Many drugs containing carboxylic acid functional groups are metabolised in vivo to ester glucuronides (1-O-acyl-beta-D-glucopyranuronates) and, of these, a number show a propensity to undergo internal isomerisation via a transacylation process, causing the carboxylic acid moiety to migrate successively to the 2-, 3- and 4-positions of the glucuronic acid. These products may be responsible, through reactions with plasma proteins, for some of the allergenic side effects in a number of non-steroidal anti-inflammatory drugs. It is important to understand those properties of the drug molecules which facilitate this reaction, and to this end we have studied the transacylation product formation and reaction kinetics in a series of aryl carboxylic acid glucuronides using NMR spectroscopy. However, the resulting 1H NMR spectra are very complex with much resonance overlap, and recourse to spectral simplification processes is necessary. Here, improvement in spectral resolution by oversampling and digital filtering to restrict the detection range of the spectrometer, thus yielding improved digital resolution, is demonstrated. The approach has been applied to the assignment of a mixture of transacylated ester glucuronides of 2-trifluoromethylbenzoic acid through the use of a two-dimensional 1H-1H TOCSY experiment.
Sigma-Aldrich
2-(Trifluoromethyl)benzoic acid, 98%
Pricing and availability is not currently available.