Merck
High affinity glutamate transport in rat cortical neurons in culture.
Molecular pharmacology (1998-01-28)
G J Wang, H J Chung, J Schnuer, K Pratt, A C Zable, M P Kavanaugh, P A Rosenberg
ABSTRACT
We assayed glutamate transport activity in cultures of rat cortical neurons containing < 0.2% astrocytes. Using [3H]L-glutamate as the tracer, sodium-dependent high affinity glutamate transport was demonstrated [K(m) = 17.2 +/- 2.4 microM; Vmax = 3.3 +/- 0.32 nmol/mg of protein/min (n = 5)]. Dihydrokainate (1 mM) inhibited uptake of radioactivity by 88 +/- 3% and had a Ki value of 65 +/- 7 microM. L-alpha-Aminoadipate (1 mM) inhibited uptake by only 25 +/- 4%. L-trans-2,4-Pyrrolidine dicarboxylate, L-serine-O-sulfate, and kainate potently inhibited transport activity with Ki values of 5.1 +/- 0.3, 56 +/- 6, and 103 +/- 9 microM, respectively (n = 3). Voltage-clamp studies of GLT1-expressing oocytes showed that, as in cortical neurons, glutamate transport was not inhibited by L-alpha-aminoadipate. Dihydrokainate was a potent inhibitor (Ki = 8 +/- 1 microM), and L-serine-O-sulfate produced a GLT1-mediated current with a K(m) value of 312 +/- 33 microM. Immunoblot analysis showed that neuronal cultures express excitatory amino acid carrier 1 (EAAC1), shown previously to be relatively insensitive to dihydrokainate, plus a trace amount of GLT1, but no GLAST. These studies establish that a major component of the glutamate transport activity of cortical neurons is dihydrokainate sensitive and distinct from the previously recognized neuronal transporter excitatory amino acid carrier 1.
Sigma-Aldrich
L-Serine O-sulfate potassium salt