Merck
  • Selective oxidation of alcohols at the benzylic position by benzeneseleninic anhydride.
Selective oxidation of alcohols at the benzylic position by benzeneseleninic anhydride.
Chemical & pharmaceutical bulletin (2004-08-12)
Naotoshi Toki, Tsuyoshi Satoh
ABSTRACT
Benzeneseleninic anhydride in the presence of tert-butyl hydroperoxide in chlorobenzene at about 70 degrees C is an effective oxidizing agent for the selective oxidation of alcohols at the benzylic position.
Sigma-Aldrich
Benzeneseleninic acid anhydride, 70%