Merck
  • Synthesis and biological evaluation of arylated novobiocin analogs as Hsp90 inhibitors.
Synthesis and biological evaluation of arylated novobiocin analogs as Hsp90 inhibitors.
Bioorganic & medicinal chemistry letters (2011-10-22)
Bhaskar Reddy Kusuma, Adam S Duerfeldt, Brian S J Blagg
ABSTRACT
Novobiocin analogs lacking labile glycosidic ether have been designed, synthesized and evaluated for Hsp90 inhibitory activity. Replacement of the synthetically complex noviose sugar with simple aromatic side chains produced analogs that maintain moderate cytotoxic activity against MCF7 and SkBR3 breast cancer cell-lines. Rationale for the preparation of des-noviose novobiocin analogs in addition to their synthesis and biological evaluation are presented herein.
Sigma-Aldrich
Novobiocin sodium salt, ≥90% (HPLC)
Sigma-Aldrich
Novobiocin sodium, meets USP testing specifications
Supelco
Novobiocin sodium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Novobiocin sodium salt, ≥93% (HPLC)