Merck
  • Evidence for the existence of terminal scandium imidos: mechanistic studies involving imido-scandium bond formation and C-H activation reactions.
Evidence for the existence of terminal scandium imidos: mechanistic studies involving imido-scandium bond formation and C-H activation reactions.
Journal of the American Chemical Society (2012-10-30)
Benjamin F Wicker, Hongjun Fan, Anne K Hickey, Marco G Crestani, Jennifer Scott, Maren Pink, Daniel J Mindiola
ABSTRACT
The anilide-methyl complex (PNP)Sc(NH[DIPP])(CH(3)) (1) [PNP(-) = bis(2-diisopropylphosphino-4-tolyl)amide, DIPP = 2,6-diisopropylphenyl] eliminates methane (k(avg) = 5.13 × 10(-4) M(-1) s(-1) at 50 °C) in the presence of pyridine to generate the transient scandium imido (PNP)Sc═N[DIPP](NC(5)H(5)) (A-py), which rapidly activates the C-H bond of pyridine in 1,2-addition fashion to form the stable pyridyl complex (PNP)Sc(NH[DIPP])(η(2)-NC(5)H(4)) (2). Mechanistic studies suggest the C-H activation process to be second order overall: first order in scandium and first order in substrate (pyridine). Pyridine binding precedes elimination of methane, and α-hydrogen abstraction is overall-rate-determining [the kinetic isotope effect (KIE) for 1-d(1) conversion to 2 was 5.37(6) at 35 °C and 4.9(14) at 50 °C] with activation parameters ΔH(‡) = 17.9(9) kcal/mol and ΔS(‡) = -18(3) cal/(mol K), consistent with an associative-type mechanism. No KIE or exchange with the anilide proton was observed when 1-d(3) was treated with pyridine or thermolyzed at 35 or 50 °C. The post-rate-determining step, C-H bond activation of pyridine, revealed a primary KIE of 1.1(2) at 35 °C for the intermolecular C-H activation reaction in pyridine versus pyridine-d(5). Complex 2 equilibrated back to the imide A-py slowly, as the isotopomer (PNP)Sc(ND[DIPP])(η(2)-NC(5)H(4)) (2-d(1)) converted to (PNP)Sc(NH[DIPP])(η(2)-NC(5)H(3)D) over 9 days at 60 °C. Molecular orbital analysis of A-py suggested that this species possesses a fairly linear scandium imido motif (169.7°) with a very short Sc-N distance of 1.84 Å. Substituted pyridines can also be activated, with the rates of C-H activation depending on both the steric and electronic properties of the substrate.
Sigma-Aldrich
Scandium, powder, 99.9% trace rare earth metals basis