- Identification of the major urinary and fecal metabolites of 3,5-dinitrobenzamide in chickens and rats.
Identification of the major urinary and fecal metabolites of 3,5-dinitrobenzamide in chickens and rats.
Chemosphere (1999-04-02)
N W Shappell, G L Larsen, J E Bakke
PMID10101847
ABSTRACT
Colostomized chickens given oral doses of 3,5-dinitrobenzamide (nitromide) cleared nitromide predominantly through the urine (58% of dose) and feces (21% of dose). Rats cleared 52% of nitromide via urinary excretion and 44% via feces. Major urinary metabolites for both chickens and rats include: 3-amino-5-nitrobenzamide, 3-acetamido-5-nitrobenzamide, 3-acetamide-5-aminobenzamide, and 3,5-diacetamidobenzamide. The major fecal metabolite in chickens was 3-acetamido-5-nitrobenzamide (67% of fecal 14C) and 3-acetamido-5-aminobenzamide in rats (approximately 50%).
MATERIALS