Merck
  • [Fibrin-depolymerization activity and the antiplatelet effect of small cyclic and linear proline-containing peptides].
[Fibrin-depolymerization activity and the antiplatelet effect of small cyclic and linear proline-containing peptides].
Izvestiia Akademii nauk. Seriia biologicheskaia (2005-06-02)
V E Pastorova, L A Liapina, I P Alshmarin, P U Ostrovskaia, T A Gudasheva, E V Lugovskoĭ
ABSTRACT
In our study of the effect of proline-containing linear di- and tri-peptides (Pro-Gly, Pro-Gly-Pro) and cyclic peptide cPro-Gly in vitro, we have found that they show depolymerizing activity towards the nonstabilized form of fibrin in the presence and in the absence of sigma-aminocapronic acid, an inhibitor of enzymatic fibrinolysis. The peptides have an inhibitory effect on platelet aggregation in platelet-enriched blood plasma, when the process is induced by ADP, they also diminish the activity of factor XIIIa in blood plasma.
Sigma-Aldrich
Pro-Gly