- Synthesis and fungicidal activity of N-2-(3-methoxy-4-propargyloxy) phenethyl amides. Part 3: stretched and heterocyclic mandelamide oomyceticides.
Synthesis and fungicidal activity of N-2-(3-methoxy-4-propargyloxy) phenethyl amides. Part 3: stretched and heterocyclic mandelamide oomyceticides.
Pest management science (2006-11-02)
Clemens Lamberth, Hans-Joachim Kempf, Miroslav Kriz
PMID17078013
ABSTRACT
Novel analogues of mandipropamid have been designed and prepared. The synthetic approach to these stretched and heterocyclic mandelamides is outlined. Biological data demonstrate their high efficacy against important plant diseases like tomato and potato late blight (Phytophthora infestans De Bary) and grape downy mildew (Plasmopara viticola Berliner & de Toni). Structure-activity relationship studies are discussed.
MATERIALS