Merck
  • One-solvent extraction of astaxanthin from lactic acid fermented shrimp wastes.
One-solvent extraction of astaxanthin from lactic acid fermented shrimp wastes.
Journal of agricultural and food chemistry (2007-11-21)
Miquel Gimeno, Jessica Yesemite Ramírez-Hernández, César Mártinez-Ibarra, Neith Pacheco, Roeb García-Arrazola, Eduardo Bárzana, Keiko Shirai
ABSTRACT
Free astaxanthin one-solvent extractions with ethanol, acetone, and liquid 1,1,1,2-tetrafluoroethane from raw and lactic acid fermented (ensilaged) shrimp residues were investigated. The total carotenoid recovery from ensilaged shrimp wastes was higher than that from non-ensilaged ones as assessed by HPLC analyses. Acetone gave the highest extraction yields of free astaxanthin with up to 115 microg/g of material. Moreover, liquid tetrafluoroethane is reported for the first time in a successful one-solvent extraction of carotenoids from shrimp.
Sigma-Aldrich
1,1,1,2-Tetrafluoroethane, ≥99%