Merck
  • HPLC-based method for determination of absolute configuration of alpha-chiral amines.
HPLC-based method for determination of absolute configuration of alpha-chiral amines.
Analytical chemistry (1993-05-15)
P A Husain, J Debnath, S W May
ABSTRACT
We introduce a novel, HPLC-based method for facile determination of the absolute configuration of alpha-chiral amines. Our method is easily applied to a variety of compounds, including amino acid derivatives. The method involves initial derivatization of the chiral amine analyte with the chiral derivatizing reagent, N-succinimidyl alpha-methoxyphenylacetate (SMPA), to produce the corresponding diastereomeric adducts. Inspection of a particular rotomer of the SMPA adduct and application of simple rules correlates absolute configuration and HPLC elution order. A key aspect of our method is that it can be used to determine absolute configuration without using enantiomeric standards of the amine analytes. Furthermore, it is of utmost significance that our method can also be used to determine absolute configuration even when only one analyte enantiomer of unknown absolute configuration is present, as is often the case for enzymatic products, naturally derived compounds, or enantiomerically enriched compounds prepared via chiral syntheses. We have observed strict adherence between predicted and observed absolute configuration for a wide variety of alpha-chiral amines. The chromatographic method we present in this paper is very practical and has several important advantages over NMR-based approaches which have been previously developed. For example, microgram quantities of an analyte in a complex enzymatic mixture can be directly analyzed by our HPLC-based method while the impurities often preclude definitive proton assignments in the NMR approach.
Product Description
Supelco
(R)-(−)-α-Methoxyphenylacetic acid, derivatization grade (chiral), LiChropur, ≥99.0%