Merck
  • Structural, electronic and magnetic properties of Cu(II) complexes of 2-substituted tropones bearing a ferrocenyl group at 5-position.
Structural, electronic and magnetic properties of Cu(II) complexes of 2-substituted tropones bearing a ferrocenyl group at 5-position.
Dalton transactions (Cambridge, England : 2003) (2010-02-18)
Tohru Nishinaga, Tomoshi Aono, Eigo Isomura, Sayaka Watanabe, Yoshihiro Miyake, Akira Miyazaki, Toshiaki Enoki, Hitoshi Miyasaka, Hiroyuki Otani, Masahiko Iyoda
ABSTRACT
Heterotrinuclear Fe(II)-Cu(II)-Fe(II) complexes [Cu(FcTropOMe)(2)(H(2)O)(2)](OTf)(2) (FcTropOMe = 5-ferrocenyl-2-methoxytropone) (1), [Cu(FcTropNEt(2))(2)](OTf)(2) (FcTropNEt(2) = 2-(N,N-diethylamino)-5-ferrocenyltropone) (2) and [Cu(FcTropNEt)(2)] (FcTropNEt = 2-(N-ethylamino)-5-ferrocenyltroponate) (3) were synthesized. In addition, a hexafluorophosphate salt of heterotrinuclear Fe(III)-Cu(II)-Fe(III) complex [Cu(FcTropNEt)(2)](2+) (3(2+)) was successfully obtained as single crystals by electrochemical oxidation of 3. By comparing the X-ray structures and absorption spectra of dicationic complexes 1 and 2, the 2-(diethylamino)tropone ligand was found to induce a greater intramolecular charge transfer (CT) from ferrocenyl to tropone-Cu(II) moieties than the 2-methoxytropone ligand. On the other hand, 3(2+) showed a broad CT band in the near-infrared (NIR) region similar to 2, which can be assigned to a transition from troponato-Cu(II) to ferrocenium moieties. As for the magnetic properties of 3(2+)(PF(6)(-))(2), measurements of temperature dependence of magnetic susceptibility and ESR on the solid state and in solution revealed the presence of a strong ferromagnetic interaction (J(Fe-Cu) = +12.0 cm(-1)) between the low spin Fe(III) ion with S = 1/2 and Cu(II) ion with S = 1/2 despite a long distance pathway via the aminotroponato and cyclopentadienyl moieties. DFT calculations supported this intramolecular ferromagnetism, which is induced by a spin polarization mechanism through the pi-spacers.
Sigma-Aldrich
Tropone, 97%