Merck
  • Development of a treatment solution for reductive dechlorination of hexachloro-1,3-butadiene in vadose zone soil.
Development of a treatment solution for reductive dechlorination of hexachloro-1,3-butadiene in vadose zone soil.
Biodegradation (2010-04-13)
Lachlan H Yee, Vibeke Aagaard, Angela Johnstone, Matthew Lee, Staffan J Kjelleberg, Mike Manefield
ABSTRACT
The biodegradation of chlorinated organics in vadose zone soils is challenging owing to the presence of oxygen, which inhibits reductive dehalogenation reactions and consequently the growth of dehalorespiring microbes. In addition, the hydraulic conductivity of vadose zone soils is typically high, hence attempts to remediate such zones with biostimulation solutions are often unsuccessful due to the short residence times for these solutions to act upon the native bacterial community. In this study we have identified sodium alginate as a hydrogel polymer that can be used to increase the residence time of a nutrient solution in an unsaturated sandy soil. Additionally we have identified neutral red as a redox active compound that can catalyse the reductive dechlorination of the chlorinated organic hexachloro-1,3-butadiene by activated sludge fed with lactate and acetate. Finally we have shown that a nutrient solution amended with neutral red and sodium alginate can lower the redox potential and reduce hexachloro-1,3-butadiene concentrations in a contaminated vadose zone soil.
Product Description
Supelco
Hexachloro-1,3-butadiene, analytical standard
Supelco
Hexachlorobutadiene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Hexachloro-1,3-butadiene, 96%