Merck
  • Gateway synthesis of daphnane congeners and their protein kinase C affinities and cell-growth activities.
Gateway synthesis of daphnane congeners and their protein kinase C affinities and cell-growth activities.
Nature chemistry (2011-07-23)
Paul A Wender, Nicole Buschmann, Nathan B Cardin, Lisa R Jones, Cindy Kan, Jung-Min Kee, John A Kowalski, Kate E Longcore
ABSTRACT
The daphnane diterpene orthoesters constitute a structurally fascinating family of natural products that exhibit a remarkable range of potent biological activities. Although partial activity information is available for some natural daphnanes, little information exists for non-natural congeners or on how changes in structure affect mode of action, function, potency or selectivity. A gateway strategy designed to provide general synthetic access to natural and non-natural daphnanes is described and utilized in the synthesis of two novel members of this class. In this study, a commercially available tartrate derivative was elaborated through a key late-stage diversification intermediate into B-ring yuanhuapin analogues to initiate exploration of the structure-function relationships of this class. Protein kinase C was identified as a cellular target for these agents, and their activity against human lung and leukaemia cell lines was evaluated. The natural product and a novel non-natural analogue exhibited significant potency, but the epimeric epoxide was essentially inactive.