Merck
  • Evaluation of sigma (σ) receptors in the antidepressant-like effects of ketamine in vitro and in vivo.
Evaluation of sigma (σ) receptors in the antidepressant-like effects of ketamine in vitro and in vivo.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology (2011-09-14)
Matthew J Robson, Meenal Elliott, Michael J Seminerio, Rae R Matsumoto
ABSTRACT
Ketamine is an NMDA antagonist and dissociative anesthetic that has been shown to display rapid acting and prolonged antidepressant activity in small-scale human clinical trials. Ketamine also binds to σ receptors, which are believed to be protein targets for a potential new class of antidepressant medications. The purpose of this study was to determine the involvement of σ receptors in the antidepressant-like actions of ketamine. Competition binding assays were performed to assess the affinity of ketamine for σ(1) and σ(2) receptors. The antidepressant-like effects of ketamine were assessed in vitro using a neurite outgrowth model and PC12 cells, and in vivo using the forced swim test. The σ receptor antagonists, NE-100 and BD1047, were evaluated in conjunction with ketamine in these assays to determine the involvement of σ receptors in the antidepressant-like effects of ketamine. Ketamine bound to both σ(1) and σ(2) receptors with μM affinities. Additionally, ketamine potentiated NGF-induced neurite outgrowth in PC12 cells and this effect was attenuated in the presence of NE-100. Ketamine also displayed antidepressant-like effects in the forced swim test; however, these effects were not attenuated by pretreatment with NE-100 or BD1047. Taken together, these data suggest that σ receptor-mediated neuronal remodeling may contribute to the antidepressant effects of ketamine.
Sigma-Aldrich
NE-100, ≥98% (HPLC)