Merck
  • Characteristics and antioxidative activity of the acetone-soluble and -insoluble fractions of a defatted rice bran extract obtained by using an aqueous organic solvent under subcritical conditions.
Characteristics and antioxidative activity of the acetone-soluble and -insoluble fractions of a defatted rice bran extract obtained by using an aqueous organic solvent under subcritical conditions.
Bioscience, biotechnology, and biochemistry (2013-03-09)
Tai-Ying Chiou, Takashi Kobayashi, Shuji Adachi
ABSTRACT
The defatted rice bran extracts obtained by being treated with various subcritical aqueous fluids at 230 °C for 5 min were further subjected to an acetone-solubilization treatment. The 1,1-diphenyl-2-picrylhydrazyl (DPPH) radical scavenging activity depended on the total phenolic content of a sample, the acetone-soluble fraction of the extract obtained with 40% (v/v) aqueous acetone showing the highest activity. Based on the UV spectrometric and HPLC analyses, the substances with higher absorbance around 280 nm and higher hydrophobicity were found solubilizing out from the extracts into the acetone during fractionation. A gel permeation chromatographic analysis suggested that the extracts comprised monomeric or oligomeric substances. The antioxidative activity was evaluated by a DPPH radical scavenging activity analysis, rancidity test, and autoxidation analysis, revealing that it would be most likely to exert a radical scavenging effect more effectively during the initial stage of lipid oxidation than during the propagation stage.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, ACS spectrophotometric grade, ≥99.5%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl