Merck
  • Structure and conformation of linear peptides. XIII. Structure of L-phenylalanyl-glycyl-glycine.
Structure and conformation of linear peptides. XIII. Structure of L-phenylalanyl-glycyl-glycine.
International journal of peptide and protein research (1989-09-01)
E Subramanian, J J Sahayamary
ABSTRACT
The crystal structure of a tripeptide, L-phenylalanyl-glycyl-glycine (C13H17N3O4), molecular weight = 279.3, has been determined. The crystals are orthorhombic, space group P2(1)2(1)2(1), with a = 5.462(1) A, b = 15.285(5), c = 16.056(4), Z = 4, and P (calc) = 1.384 g.cm-3. The final R-index is 0.052 for 866 reflections with sin theta/lambda less than or equal to 0.55 A-1 and I greater than 1 sigma. The molecule exists as a zwitterion, with the N-terminus protonated and the C-terminus in an ionized form. Both the peptide units are in the trans configuration and planar, though one of them shows significant deviations from planarity ([delta w[ = 5.1 degrees). The peptide backbone is folded, with the torsion angles of: psi 1 = 116.2(5) degrees, omega 1 = 178.8(4), phi 2 = -89.7(5). psi 2 = -28.9(6), omega 2 = -174.9(4), phi 3 = 134.9(5), psi 31 = 7.8(6), psi 32 = -172.6(4). The terminal glycine adopts a "D-residue" conformation. For the sidechain of phenylalanine, chi 1 = 175.5(4), chi 2 = -127.0(6).
Sigma-Aldrich
Phe-Gly-Gly
Pricing and availability is not currently available.