Merck
  • Maternal hepatic and embryonic effects of 1,2,3,4-tetrachlorobenzene in the rat.
Maternal hepatic and embryonic effects of 1,2,3,4-tetrachlorobenzene in the rat.
Toxicology (1983-03-01)
K T Kitchin, M T Ebron
ABSTRACT
To assess possible maternal hepatic and reproductive effects of this uncharged, low molecular weight, lipophilic chlorinated benzene 0, 100, 300 and 1000 mg/kg/day of 1,2,3,4-tetrachlorobenzene (TCB) was orally administered to pregnant rats on days 9-13 of gestation and the animals were killed on day 14 of pregnancy. Phenobarbital and beta-naphthoflavone were administered to other pregnant rats as positive hepatic controls. Maternal mortality (7/19 rats) was increased and body weight gain was greatly decreased in the 1000 mg/kg/day TCB group. Liver to body weight ratio and hepatic microsomal protein content were unaffected by any TCB treatment. On day 14 maternal NADPH-cytochrome c reductase activity was increased at 1000 mg/kg/day, while the maternal hepatic microsomal cytochrome P-450 content was significantly induced by both 300 and 1000 mg/kg/day of TCB. Microsomal N-demethylation of aminopyrine was increased from 2.6 to 4.0 and 4.5 nmol/mg protein/min at doses of 300 and 1000 mg/kg TCB, respectively. However, maternal hepatic microsomal ethoxyresorufin O-deethylase activity was not consistently increased by TCB. Hepatic glutathione S-transferase activity towards 1,2-dichloro-4-nitrobenzene was increased only by the 1000 mg/kg/day TCB treatment. The rate of microsomal p-nitrophenol and phenolphthalein glucuronidation was increased by TCB administration. Embryonic growth was adversely affected by TCB treatment. Yolk sac diameter, embryonic crown-rump length, and head length were all decreased by treatment with 300 mg/kg/day TCB. This TCB treatment did not significantly elevate the number of dead or abnormal embryos.
Sigma-Aldrich
1,2,3,4-Tetrachlorobenzene, 98%
Supelco
1,2,3,4-Tetrachlorobenzene, PESTANAL®, analytical standard