Merck
  • Phosphonopeptide antibacterial agents related to alafosfalin: design, synthesis, and structure-activity relationships.
Phosphonopeptide antibacterial agents related to alafosfalin: design, synthesis, and structure-activity relationships.
Antimicrobial agents and chemotherapy (1980-12-01)
F R Atherton, M J Hall, C H Hassall, S W Holmes, R W Lambert, W J Lloyd, P S Ringrose
ABSTRACT
Dipeptide variants of alafosfalin (L-alanyl-L-1-aminoethylphosphonic acid) with substantial differences in potency and antibacterial spectrum in vitro and in vivo have been synthesized. Certain dipeptides with alternatives to the L-alanyl residue had broader antibacterial spectra; activity against Pseudomonas aeruginosa was included. Some compounds had better in vivo activity than alafosfalin when introduced into infected rodents orally, but for the majority of the more active phosphonodipeptides, parenteral administration was more effective. Certain oligopeptides derived from the more active phosphonodipeptides possessed good in vitro activity against an extended range of organisms; they included Haemophilus influenzae, Streptococcus faecalis, and Streptococcus pneumoniae. The in vivo activity of some of these phosphono-oligopeptides was significantly greater than that of the parent dipeptide and correlated well with the in vitro results. This indicates that phosphono-oligopeptides exert part of their in vivo action directly, in addition to that arising from smaller peptides produced by peptidase cleavage.
Sigma-Aldrich
L-Alanyl-L-1-aminoethylphosphonic acid, ≥95% (HPLC)