Merck
  • Influence of bromfenvinfos alone and in mixture with methoxychlor on the blood indices of laboratory mice.
Influence of bromfenvinfos alone and in mixture with methoxychlor on the blood indices of laboratory mice.
Polish journal of pharmacology and pharmacy (1982-01-01)
K I Zaleska-Freljan, B Kosicka
ABSTRACT
Bromfenvinfos (O,O-diethyl-O-1-(2,4-dichlorphenyl)-2-bromvinyl phosphate, BrV, 12.23 mg/kg/day) alone and in combination with methoxychlor (1,1,1-trichlor-2,2-(4-methoxyphenyl) ethane, MeOCl, 24.66 mg/kg/day) were administered per os daily for 6 weeks to male and female laboratory mice in the olive-oil suspension. Total leukocyte and erythrocyte count, hemoglobin level, the percentage of leukocytes and the plasma proteins were determined. BrV and BrV + MeOCl depressed the blood hemoglobin level; BrV and BrV + MeOCl treatment reduced also the erythrocyte diameter.
Product Description
Supelco
Bromfenvinphos-ethyl, PESTANAL®, analytical standard