Merck
  • Quantitation of the anticonvulsant cinromide (3-bromo-N-ethylcinnamamide) and its major plasma metabolites by thin-layer chromatography.
Quantitation of the anticonvulsant cinromide (3-bromo-N-ethylcinnamamide) and its major plasma metabolites by thin-layer chromatography.
Journal of chromatography (1980-12-12)
R L DeAngelis, M M Robinson, A R Brown, T E Johnson, R M Welch
ABSTRACT
A quantitative thin-layer chromatography (TLC) procedure is described for the analysis of cinromide (3-bromo-N-ethylcinnamamide) and its two major metabolites, 3-bromocinnamamide and 3-bromocinnamic acid in plasma of the dog. These compounds were recovered from acidified plasma by extraction into benzene with a recovery of 95 +/- 5%. All three compounds were quantitated directly on a TLC plate by ultraviolet absorbance densitometry at 270 nm. The linear dynamic range for the quantitation of the compounds on a TLC plate ranged between 10 and 1000 ng. The complete procedure is useful in the working range of 50 ng/ml to 100 microgram/ml of plasma with a coefficient of variability of about 10%. Specificity of the method for parent drug and each of its plasma metabolites was confirmed by high-performance liquid chromatography. The method was used to determine the pharmacokinetics of cinromide and its two major plasma metabolites in dogs following a single oral dose of the drug.
Sigma-Aldrich
3-Bromocinnamic acid, predominantly trans, 98%