Merck
  • Identification of hepatic metabolites of two highly carcinogenic polycyclic aza-aromatic compounds, 7,9-dimethylbenz[c]acridine and 7,10-dimethylbenz[c]acridine.
Identification of hepatic metabolites of two highly carcinogenic polycyclic aza-aromatic compounds, 7,9-dimethylbenz[c]acridine and 7,10-dimethylbenz[c]acridine.
Chemical research in toxicology (1995-03-01)
Y Ye, C C Duke, G M Holder
ABSTRACT
The hepatic microsomal metabolites of the highly carcinogenic dimethylbenzacridines, 7,9-dimethylbenz[c]acridine (7,9-DMBAC), and 7,10-dimethylbenz[c]acridine (7,10-DMBAC) were obtained with preparations from 3-methylcholanthrene-pretreated rats. Metabolites were separated by reversed-phase HPLC and characterized using UV spectral data and chemical ionization-mass spectrometry after trimethylsilylation and GC. Comparisons with products formed in the presence of the epoxide hydrolase inhibitor, 1,1,1-trichloropropane 2,3-oxide and with those formed from the three synthetic alcohol derivatives of each parent compound, aided the assignment of firm or tentative structures to 16 products from 7,9-DMBAC found in 22 reversed-phase chromatographic peaks, and for 17 products of 7,10-DMBAC found in 19 chromatographic peaks. The more abundant metabolites were derived from oxidation of the methyl groups. Other metabolites were dihydrodiols, epoxides, phenols and secondary metabolites. The 9-methyl group prevented dihydrodiol formation at the 8,9-position from 7,9-DMBAC, and for each carcinogen, the 3,4-dihydrodiol was formed. As well, 3,4-dihydrodiols of methyl oxidized compounds were found.
Sigma-Aldrich
7,9-Dimethylbenz[c]acridine, technical grade