Merck
  • Amine-reactive forms of a luminescent diethylenetriaminepentaacetic acid chelate of terbium and europium: attachment to DNA and energy transfer measurements.
Amine-reactive forms of a luminescent diethylenetriaminepentaacetic acid chelate of terbium and europium: attachment to DNA and energy transfer measurements.
Bioconjugate chemistry (1997-03-01)
M Li, P R Selvin
ABSTRACT
An isothiocyanate form of a lanthanide chelate which is highly luminescent when bound to terbium or europium has been synthesized. The chelate consists of diethylenetriaminepentaacetic acid (DTPA) covalently joined to a chromophore, 7-amino-4-methyl-2(1H)-quinolinone (cs124), and to L-p-aminophenylalanine, in which the aromatic amine was further converted to an isothiocyanate group. Ethylenediamine was also used in place of aminophenylalanine, but the isothiocyanate formed from the aliphatic amine was significantly less reactive. Site-specific attachments to triglycine and to the 5' ends of amine-modified DNA oligomers have been made. In addition, as an alternative method of coupling to macromolecules, DTPA anhydride-cs124 can be used to react specifically with a 5' amine group on base-deprotected synthetic DNA oligomers. Synthesis and purification is relatively straightforward in both cases, and luminescent properties are favorable for several applications, including as nonisotopic labels, as long-lifetime alternatives to fluorophores in imaging and diagnostics and particularly as donors in luminescence resonance energy transfer. Energy transfer measurements are consistent with previously reported measurements using different attachment mechanisms.
Sigma-Aldrich
Carbostyril 124, 99%