Merck
  • Readily synthesised arylamino fumaronitrile for non-doped red organic light-emitting diodes.
Readily synthesised arylamino fumaronitrile for non-doped red organic light-emitting diodes.
Chemical communications (Cambridge, England) (2003-11-05)
Hsiu-Chih Yeh, Shi-Jay Yeh, Chin-Ti Chen
ABSTRACT
Bright (maximum 10034 cd m(-2), 455 cd m(-2) at 20 mA cm(-2)) and efficient (maximum 2.4% at 4 mA cm(-2)) red (lambda(max)el 634-636 nm) organic light-emitting diodes employ arylamino-substituted fumaronitrile as the novel host emitter, which is readily prepared and easily purified.
Sigma-Aldrich
4-Bromophenylacetonitrile, 98%