Merck
  • Synthesis and evaluation of L-glutamic Acid analogs as potential anticancer agents.
Synthesis and evaluation of L-glutamic Acid analogs as potential anticancer agents.
Indian journal of pharmaceutical sciences (2008-03-01)
C L Vishwanathan, S Deb, A Jain, T Lokhande, Aarti Juvekar
ABSTRACT
Four N-(benzenesulfonyl)-L-glutamic acid bis(p-substituted phenylhydrazides) were synthesized and evaluated for anticancer activity in vitro in DU-145 and PC-3 prostate cancer and in COLO-205 colon cancer cell lines by MTT assay. The analog with the nitro group substitution exhibited potent activity (% Inhibition 84.7 and 72.0 in DU-145 and PC-3 respectively at 80 mug/ml concentration). Another series of substituted 1-(benzenesulfonyl)-5-oxopyrrolidine 2-carboxamides (11a-f) were synthesized and evaluated for anticancer activity in vitro in colon (COLO-205), breast (Zr-75-1) and prostate (PC-3) cancer cell lines by MTT assay using adriamycin as standard. Test compounds 11a-c showed potent activity (% Inhibition 61.2 to 79.2 at 20 mug/ml and 67.2 to 87.2 at 40 mug/ml) in PC-3 cell line which is superior to the activity of Adriamycin. In comparison compounds 11d-f were less potent. In Zr-75-1 cell line 11a-e showed % inhibition ranging from 32.4 to 54.9 at 10 mug/ml concentration while in COLO-205 cell line 11a-f showed poor activity.
Product Description
ZR-75-1, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 87012601, human breast, Epithelial