Merck
  • Prolonged incubation in calcium chloride improves the competence of Escherichia coli cells.
Prolonged incubation in calcium chloride improves the competence of Escherichia coli cells.
Gene (1979-05-01)
M Dagert, S D Ehrlich
PMID383576
ABSTRACT
Escherichia coli cells are 4--6 times more transformable and 20--30 times more competent after 24 h incubation in cold calcium chloride than immediately after calcium chloride treatment. With 24-h-old competent cells we obtained routinely 2 . 10(7) transformants per microgram of pBR322 DNA, and transformed over 20% of viable cells.
Sigma-Aldrich
Calcium chloride hexahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride hexahydrate, meets analytical specification of Ph. Eur., E509, 97-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.