Merck
  • Effect of thiamine hydrochloride, pyridoxine hydrochloride and calcium-d-pantothenate on the patulin content of apple juice concentrate.
Effect of thiamine hydrochloride, pyridoxine hydrochloride and calcium-d-pantothenate on the patulin content of apple juice concentrate.
Die Nahrung (2002-09-13)
Serafettin Yazici, Y Sedat Velioglu
ABSTRACT
Thiamine hydrochloride, pyridoxine hydrochloride and calcium-d-pantothenate were applied apple juice concentrates (AJC) at various doses in order to reduce the patulin content. AJC samples containing high levels of patulin were stored at 22 +/- 2 degrees C and 4 degrees C for 6 months after vitamins were added. Patulin was fully degraded at the end of a 6-month period in samples stored at 22 +/- 2 degrees C, on the other hand, other quality parameters diminished significantly. Without any considerable reduction on other quality parameters, applications of 1000 and 2500 mg/kg calcium-d-pantothenate resulted in reduction of patulin of 73.6 and 94.3%, respectively, however, 42.1% of patulin reduction was observed in the control sample of AJC stored for 1 month at 22 +/- 2 degrees C. Addition of thiamine hydrochloride (1000 mg/kg), pyrodoxine hydrochloride (625 or 875 mg/kg) and calcium-d-pantothenate (1000 or 2500 mg/kg) into the samples and storage at 4 degrees C for 6 months yielded 55.5 to 67.7% of patulin reduction which was only 35.8% for the control while the other quality parameters were protected adequately.
Sigma-Aldrich
Thiamine hydrochloride, tested according to Ph. Eur.
Pricing and availability is not currently available.