Merck
  • Lasting chemiluminescence of 3-indoleglyoxylyl chloride and its enhancement.
Lasting chemiluminescence of 3-indoleglyoxylyl chloride and its enhancement.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2003-02-01)
Manabu Nakazono, Yumiko Sho, Kiyoshi Zaitsu
ABSTRACT
The chemiluminescence (CL) intensities of various indole derivatives substituted with a glyoxylyl group at the 3-position and a hydroxyl group at the 5-position of the indole ring were compared upon the addition of H2O2 in alkaline media. The CL intensities of 3-indoleglyoxylyl chloride, 3-indoleglyoxylic acid, 5-hydroxyindole and 5-benzyloxyindole in CH3CN were 5.9-, 48-, 5.9- and 3.3-fold stronger than that of 3-methylindole. A lasting CL of 3-indoleglyoxylyl chloride was found. Under appropriate conditions, the CL emission reached a maximum within 10 min after the addition of H2O2 in the presence of NaOH, and the intensity was retained for 25 min. One of the final products via the CL reaction of 3-indoleglyoxylyl chloride was indole-3-carboxylic acid. 3-Indoleglyoxylyl chloride emitted light by decompositions via both dioxetane and dioxetanedione. An enhancement effect of beta-cyclodextrin and bovine serum albumin on the CL of 3-indoleglyoxylyl chloride was also found.
Sigma-Aldrich
3-Indoleglyoxylic acid, 98%
Sigma-Aldrich
Methyl 3-indoleglyoxylate, 98%
Sigma-Aldrich
3-Indoleglyoxylyl chloride, 98%