Merck
  • Highly enantioselective catalytic phenylation of ketones with a constrained geometry titanium catalyst.
Highly enantioselective catalytic phenylation of ketones with a constrained geometry titanium catalyst.
Organic letters (2003-09-26)
Celina García, Patrick J Walsh
ABSTRACT
[reaction: see text] The catalytic asymmetric addition of phenyl groups from diphenylzinc to ketones is reported. The catalyst, generated from a dihydroxy bis(sulfonamide) ligand and titanium tetraisopropoxide, gives good to excellent enantioselectivities with a range of substrates.
Sigma-Aldrich
3′-Chloropropiophenone, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
3′-(Trifluoromethyl)acetophenone, 99%
Pricing and availability is not currently available.