Merck
Efficacy of flutrimazole 1% powder in the treatment of tinea pedis.
Mycoses (2003-07-23)
J Pereda, X Noguera, E Boncompte, M Algueró, I Izquierdo
ABSTRACT
The aim of the study was to compare the efficacy and tolerability of flutrimazole 1% powder vs. bifonazole 1% powder in treating tinea pedis. A multicentre, double blind, randomized, parallel and comparative study was conducted. Two hundred and twenty-two patients with clinically and mycologically confirmed tinea pedis were randomized to flutrimazole (n = 136) or bifonazole (n = 138) 1% powder applied twice daily for 4 weeks. The corresponding clinical cure rates were assessed at 2 and 4 weeks of treatment, and the global (clinical and mycological) cure rates were determined at the fourth week. Clinical cure rates were 83.5 and 82.4% for flutrimazole and bifonazole, respectively (95% CI: -0.0806 to 0.1009). Global cure rates were observed in 65.3 and 70.1% of patients treated with flutrimazole and bifonazole, respectively (95% CI: -0.0828 to 0.1779). Three non serious adverse events at the application site--itching (one patient per group) and dishydrotic eczema (one patient treated with flutrimazole)--were recorded during the study. These results support that flutrimazol 1% powder applied twice daily for a duration of 4 weeks is highly effective in the treatment of tinea pedis, showing a similar therapeutic profile with that of bifonazole 1% powder.